Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCTTCTGTAACGTACCTGACAGT[A/G]ATTTCAGGCAGAACTGCCGGGTATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RWDD2B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RWDD2B - RWD domain containing 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320724.1 | 546 | Silent Mutation | ATC,ATT | I,I 89 | NP_001307653.1 | |
NM_016940.2 | 546 | Silent Mutation | ATC,ATT | I,I 118 | NP_058636.1 |