Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCATCTAAGTTCTGATTTTTGGA[A/G]CAAGAGGAAAAGGAGTAGGAGAAAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C21orf62 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C21orf62 - chromosome 21 open reading frame 62 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162495.2 | 2198 | UTR 3 | NP_001155967.2 | |||
NM_001162496.2 | 2198 | UTR 3 | NP_001155968.2 | |||
NM_019596.5 | 2198 | UTR 3 | NP_062542.5 | |||
XM_011529653.2 | 2198 | Intron | XP_011527955.1 |
C21orf62-AS1 - C21orf62 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |