Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTATTGGTTATTTACTTTTGGAC[C/T]TCTCACCTTCCGGGATAATTCTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608369 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALNT13 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GALNT13 - polypeptide N-acetylgalactosaminyltransferase 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301627.1 | Intron | NP_001288556.1 | ||||
NM_052917.3 | Intron | NP_443149.2 | ||||
XM_011510537.2 | Intron | XP_011508839.1 | ||||
XM_011510538.2 | Intron | XP_011508840.1 | ||||
XM_017003258.1 | Intron | XP_016858747.1 | ||||
XM_017003259.1 | Intron | XP_016858748.1 | ||||
XM_017003260.1 | Intron | XP_016858749.1 | ||||
XM_017003261.1 | Intron | XP_016858750.1 | ||||
XM_017003262.1 | Intron | XP_016858751.1 | ||||
XM_017003263.1 | Intron | XP_016858752.1 |
Set Membership: |
HapMap |