Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAAATACTTCCTCAATAGCAGGG[A/G]AAAAAAAGCAAGAGAAAGACTGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616285 MIM: 612941 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FMNL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FMNL2 - formin like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRPF40A - pre-mRNA processing factor 40 homolog A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017892.3 | 3876 | UTR 3 | NP_060362.3 | |||
XM_005246676.4 | 3876 | UTR 3 | XP_005246733.3 | |||
XM_005246677.4 | 3876 | Intron | XP_005246734.2 | |||
XM_011511446.2 | 3876 | UTR 3 | XP_011509748.1 | |||
XM_011511447.2 | 3876 | Intron | XP_011509749.1 | |||
XM_011511448.2 | 3876 | UTR 3 | XP_011509750.1 | |||
XM_011511449.2 | 3876 | UTR 3 | XP_011509751.1 | |||
XM_017004453.1 | 3876 | UTR 3 | XP_016859942.1 | |||
XM_017004454.1 | 3876 | UTR 3 | XP_016859943.1 | |||
XM_017004455.1 | 3876 | UTR 3 | XP_016859944.1 | |||
XM_017004456.1 | 3876 | UTR 3 | XP_016859945.1 | |||
XM_017004457.1 | 3876 | UTR 3 | XP_016859946.1 | |||
XM_017004458.1 | 3876 | UTR 3 | XP_016859947.1 | |||
XM_017004459.1 | 3876 | UTR 3 | XP_016859948.1 | |||
XM_017004460.1 | 3876 | UTR 3 | XP_016859949.1 | |||
XM_017004461.1 | 3876 | UTR 3 | XP_016859950.1 |
Set Membership: |
HapMap |