Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACAGTCTCCAATGTCATCAGCTC[A/C]ATTGTCTTTGGGGACCGCTTTGACT
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608055 | ||||||||||||||||||||||||||||||||
Literature Links: |
CYP2A13 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
CYP2A13*1K,g.1991C>A | ||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian
|
CEPH (CEU) - Not Available | |||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | |||
SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CYP2A13 - cytochrome P450 family 2 subfamily A member 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000766.4 | Intron | NP_000757.2 |
Set Membership: |
DME Validated Inventoried |