Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATCAAGAATTATGTTTTACCTCA[A/G]CGGAGCTTCTTGGATCATGAATTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
48 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606756 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSD17B7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HSD17B7 - hydroxysteroid 17-beta dehydrogenase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304512.1 | Intron | NP_001291441.1 | ||||
NM_001304513.1 | Intron | NP_001291442.1 | ||||
NM_016371.3 | Intron | NP_057455.1 |
Set Membership: |
HapMap |