Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGTGCTTACAGGCTCCCTGCCGA[A/G]TCGCAACTGTTTACAGGGCTCAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 136130 | ||||||||||||||||||||
Literature Links: |
FMO1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FMO1 - flavin containing monooxygenase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282692.1 | Intron | NP_001269621.1 | ||||
NM_001282693.1 | Intron | NP_001269622.1 | ||||
NM_001282694.1 | Intron | NP_001269623.1 | ||||
NM_002021.2 | Intron | NP_002012.1 | ||||
XM_005245037.3 | Intron | XP_005245094.1 | ||||
XM_005245038.3 | Intron | XP_005245095.1 | ||||
XM_006711241.3 | Intron | XP_006711304.1 | ||||
XM_006711242.3 | Intron | XP_006711305.1 |