Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAGTGTCCCAGTGGGGGATGTGG[C/T]TGTGGTACCTACTGAAGGAAATGAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604409 | |||||||||||||||||||||||
Literature Links: |
GMEB1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
GMEB1 - glucocorticoid modulatory element binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319674.1 | 286 | Missense Mutation | GCT,GTT | A,V 14 | NP_001306603.1 | |
NM_006582.3 | 286 | Missense Mutation | GCT,GTT | A,V 14 | NP_006573.2 | |
NM_024482.2 | 286 | Missense Mutation | GCT,GTT | A,V 14 | NP_077808.1 | |
XM_011540518.2 | 286 | Missense Mutation | GCT,GTT | A,V 14 | XP_011538820.1 | |
XM_011540519.2 | 286 | Missense Mutation | GCT,GTT | A,V 14 | XP_011538821.1 | |
XM_011540521.2 | 286 | Missense Mutation | GCT,GTT | A,V 14 | XP_011538823.1 | |
XM_011540522.2 | 286 | UTR 5 | XP_011538824.1 | |||
XM_017000087.1 | 286 | Missense Mutation | GCT,GTT | A,V 14 | XP_016855576.1 |