Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCATGGATTTTGATATACGGGGG[G/T]GGAGGGTTAGTTCTAGGACTACCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 612106 | ||||||||||||||||||||
Literature Links: |
ZBTB40 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZBTB40 - zinc finger and BTB domain containing 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083621.1 | Intron | NP_001077090.1 | ||||
NM_014870.3 | Intron | NP_055685.3 | ||||
XM_011542499.2 | Intron | XP_011540801.1 | ||||
XM_011542500.2 | Intron | XP_011540802.1 | ||||
XM_017003003.1 | Intron | XP_016858492.1 |