Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGGTGATTGGGAACTTGACTGGG[A/C]CTTCTAAACTACTCAACAAAACTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
48 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDR64 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
WDR64 - WD repeat domain 64 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144625.4 | Intron | NP_653226.4 | ||||
XM_006711736.3 | Intron | XP_006711799.1 | ||||
XM_011544085.2 | Intron | XP_011542387.1 | ||||
XM_011544086.2 | Intron | XP_011542388.1 | ||||
XM_011544087.2 | Intron | XP_011542389.1 | ||||
XM_011544091.1 | Intron | XP_011542393.1 | ||||
XM_011544092.2 | Intron | XP_011542394.1 | ||||
XM_017000315.1 | Intron | XP_016855804.1 |