Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610073 MIM: 600258 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ORMDL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ORMDL1 - ORMDL sphingolipid biosynthesis regulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128150.1 | 30 | UTR 5 | NP_001121622.1 | |||
NM_016467.4 | 30 | UTR 5 | NP_057551.1 | |||
XM_005246970.2 | 30 | Intron | XP_005247027.1 | |||
XM_011512199.2 | 30 | UTR 5 | XP_011510501.1 | |||
XM_011512200.2 | 30 | Intron | XP_011510502.1 | |||
XM_017005347.1 | 30 | Intron | XP_016860836.1 | |||
XM_017005348.1 | 30 | Intron | XP_016860837.1 |
Set Membership: |
HapMap |