Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610073 MIM: 600258 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ORMDL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ORMDL1 - ORMDL sphingolipid biosynthesis regulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128150.1 | Intron | NP_001121622.1 | ||||
NM_016467.4 | Intron | NP_057551.1 | ||||
XM_005246970.2 | Intron | XP_005247027.1 | ||||
XM_011512199.2 | Intron | XP_011510501.1 | ||||
XM_011512200.2 | Intron | XP_011510502.1 | ||||
XM_017005347.1 | Intron | XP_016860836.1 | ||||
XM_017005348.1 | Intron | XP_016860837.1 |
PMS1 - PMS1 homolog 1, mismatch repair system component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000534.4 | Intron | NP_000525.1 | ||||
NM_001128143.1 | Intron | NP_001121615.1 | ||||
NM_001128144.1 | Intron | NP_001121616.1 | ||||
NM_001289408.1 | Intron | NP_001276337.1 | ||||
NM_001289409.1 | Intron | NP_001276338.1 | ||||
NM_001321044.1 | Intron | NP_001307973.1 | ||||
NM_001321045.1 | Intron | NP_001307974.1 | ||||
NM_001321046.1 | Intron | NP_001307975.1 | ||||
NM_001321047.1 | Intron | NP_001307976.1 | ||||
NM_001321048.1 | Intron | NP_001307977.1 | ||||
NM_001321049.1 | Intron | NP_001307978.1 | ||||
NM_001321051.1 | Intron | NP_001307980.1 | ||||
XM_006712596.1 | Intron | XP_006712659.1 | ||||
XM_011511356.2 | Intron | XP_011509658.1 | ||||
XM_017004344.1 | Intron | XP_016859833.1 | ||||
XM_017004345.1 | Intron | XP_016859834.1 | ||||
XM_017004346.1 | Intron | XP_016859835.1 | ||||
XM_017004347.1 | Intron | XP_016859836.1 | ||||
XM_017004348.1 | Intron | XP_016859837.1 | ||||
XM_017004349.1 | Intron | XP_016859838.1 | ||||
XM_017004350.1 | Intron | XP_016859839.1 | ||||
XM_017004351.1 | Intron | XP_016859840.1 |