Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGATTTCACTTCTCGATACTGGCC[A/G]TGCTACTGCCTTCTTGACACTGCCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603835 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDUFA10 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
NDUFA10 - NADH:ubiquinone oxidoreductase subunit A10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322019.1 | Intron | NP_001308948.1 | ||||
NM_001322020.1 | Intron | NP_001308949.1 | ||||
NM_004544.3 | Intron | NP_004535.1 | ||||
XM_011511228.2 | Intron | XP_011509530.1 |
Set Membership: |
HapMap |