Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCTCGGCCAATCTGGGCGGCGT[C/G]CCCAGCAAGAGATTAAAGATGCAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607912 | ||||||||||||||||||||
Literature Links: |
SELT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SELT - selenoprotein T | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016275.3 | 162 | Silent Mutation | NP_057359.2 |
SERP1 - stress-associated endoplasmic reticulum protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014445.3 | 162 | Intron | NP_055260.1 | |||
XM_011512698.2 | 162 | Intron | XP_011511000.1 |