Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
GPR160 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese)
|
|||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
GPR160 - G protein-coupled receptor 160 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PHC3 - polyhomeotic homolog 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308116.1 | 8367 | UTR 3 | NP_001295045.1 | |||
NM_024947.3 | 8367 | UTR 3 | NP_079223.3 | |||
XM_005247789.2 | 8367 | UTR 3 | XP_005247846.1 | |||
XM_005247791.3 | 8367 | Intron | XP_005247848.1 | |||
XM_005247793.2 | 8367 | Intron | XP_005247850.1 | |||
XM_006713753.1 | 8367 | UTR 3 | XP_006713816.1 | |||
XM_006713754.3 | 8367 | UTR 3 | XP_006713817.1 | |||
XM_006713756.2 | 8367 | UTR 3 | XP_006713819.1 | |||
XM_006713757.3 | 8367 | Intron | XP_006713820.1 | |||
XM_011513186.2 | 8367 | UTR 3 | XP_011511488.1 | |||
XM_011513187.2 | 8367 | UTR 3 | XP_011511489.1 | |||
XM_011513188.2 | 8367 | UTR 3 | XP_011511490.1 | |||
XM_011513189.2 | 8367 | Intron | XP_011511491.1 | |||
XM_017007235.1 | 8367 | UTR 3 | XP_016862724.1 | |||
XM_017007236.1 | 8367 | UTR 3 | XP_016862725.1 | |||
XM_017007237.1 | 8367 | UTR 3 | XP_016862726.1 | |||
XM_017007238.1 | 8367 | UTR 3 | XP_016862727.1 | |||
XM_017007239.1 | 8367 | UTR 3 | XP_016862728.1 |
Set Membership: |
HapMap |