Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147183 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBPJ PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RBPJ - recombination signal binding protein for immunoglobulin kappa J region | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005349.3 | Intron | NP_005340.2 | ||||
NM_015874.4 | Intron | NP_056958.3 | ||||
NM_203283.2 | Intron | NP_976028.1 | ||||
NM_203284.2 | Intron | NP_976029.1 | ||||
XM_005248161.3 | Intron | XP_005248218.1 | ||||
XM_011513840.2 | Intron | XP_011512142.1 | ||||
XM_017008170.1 | Intron | XP_016863659.1 | ||||
XM_017008171.1 | Intron | XP_016863660.1 | ||||
XM_017008172.1 | Intron | XP_016863661.1 | ||||
XM_017008173.1 | Intron | XP_016863662.1 | ||||
XM_017008174.1 | Intron | XP_016863663.1 | ||||
XM_017008175.1 | Intron | XP_016863664.1 |
Set Membership: |
HapMap |