Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616344 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTC23L PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TTC23L - tetratricopeptide repeat domain 23 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317949.1 | 483 | Missense Mutation | AAA,GAA | K,E 76 | NP_001304878.1 | |
NM_144725.3 | 483 | Missense Mutation | AAA,GAA | K,E 67 | NP_653326.3 | |
XM_011513984.2 | 483 | Missense Mutation | AAA,GAA | K,E 67 | XP_011512286.1 | |
XM_011513985.2 | 483 | Missense Mutation | AAA,GAA | K,E 67 | XP_011512287.1 | |
XM_011513987.2 | 483 | Missense Mutation | AAA,GAA | K,E 67 | XP_011512289.1 | |
XM_017009119.1 | 483 | Missense Mutation | AAA,GAA | K,E 76 | XP_016864608.1 | |
XM_017009120.1 | 483 | Missense Mutation | AAA,GAA | K,E 67 | XP_016864609.1 | |
XM_017009121.1 | 483 | Missense Mutation | AAA,GAA | K,E 67 | XP_016864610.1 | |
XM_017009122.1 | 483 | Missense Mutation | AAA,GAA | K,E 67 | XP_016864611.1 | |
XM_017009123.1 | 483 | Missense Mutation | AAA,GAA | K,E 67 | XP_016864612.1 | |
XM_017009124.1 | 483 | Intron | XP_016864613.1 | |||
XM_017009125.1 | 483 | Intron | XP_016864614.1 | |||
XM_017009126.1 | 483 | Intron | XP_016864615.1 |