Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602851 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRV1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADGRV1 - adhesion G protein-coupled receptor V1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032119.3 | Intron | NP_115495.3 | ||||
XM_017009963.1 | Intron | XP_016865452.1 | ||||
XM_017009964.1 | Intron | XP_016865453.1 | ||||
XM_017009965.1 | Intron | XP_016865454.1 | ||||
XM_017009966.1 | Intron | XP_016865455.1 | ||||
XM_017009967.1 | Intron | XP_016865456.1 | ||||
XM_017009968.1 | Intron | XP_016865457.1 | ||||
XM_017009969.1 | Intron | XP_016865458.1 | ||||
XM_017009970.1 | Intron | XP_016865459.1 | ||||
XM_017009971.1 | Intron | XP_016865460.1 | ||||
XM_017009972.1 | Intron | XP_016865461.1 | ||||
XM_017009973.1 | Intron | XP_016865462.1 | ||||
XM_017009974.1 | Intron | XP_016865463.1 |