Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCTTAGTGGAGTGGGATATGCTC[C/T]AGAGAAGCAGCAGCCCATGCACTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 118850 | ||||||||||||||||||||
Literature Links: |
CGA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CGA - glycoprotein hormones, alpha polypeptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000735.3 | 410 | Missense Mutation | AGA,GGA | R,G 59 | NP_000726.1 | |
NM_001252383.1 | 410 | Missense Mutation | AGA,GGA | R,G 90 | NP_001239312.1 |