Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAAAGGCAAAGATGCTAAACTGC[C/T]AATAACAATAAGGAATAGCTGTATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608966 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DACT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DACT2 - dishevelled binding antagonist of beta catenin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286350.1 | Intron | NP_001273279.1 | ||||
NM_001286351.1 | Intron | NP_001273280.1 | ||||
NM_214462.4 | Intron | NP_999627.2 | ||||
XM_011535507.2 | Intron | XP_011533809.1 |