Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602425 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP3K4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP3K4 - mitogen-activated protein kinase kinase kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291958.1 | Intron | NP_001278887.1 | ||||
NM_001301072.1 | Intron | NP_001288001.1 | ||||
NM_005922.3 | Intron | NP_005913.2 | ||||
NM_006724.3 | Intron | NP_006715.2 | ||||
XM_005266989.2 | Intron | XP_005267046.1 | ||||
XM_017010869.1 | Intron | XP_016866358.1 |