Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATCATTGATGGCAGCAATGGGAA[G/T]ACCCCAGTTGGCTACTGGGCCCCAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614738 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MPC1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MPC1 - mitochondrial pyruvate carrier 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270879.1 | 1026 | UTR 5 | NP_001257808.1 | |||
NM_016098.3 | 1026 | Missense Mutation | ATT,CTT | I,L 36 | NP_057182.1 | |
XM_011535894.1 | 1026 | UTR 5 | XP_011534196.1 | |||
XM_011535895.1 | 1026 | UTR 5 | XP_011534197.1 |