Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609149 | ||||||||||||||||||||
Literature Links: |
SLC29A4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC29A4 - solute carrier family 29 member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNRC18 - trinucleotide repeat containing 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080495.2 | 12440 | UTR 3 | NP_001073964.2 | |||
XM_017012728.1 | 12440 | UTR 3 | XP_016868217.1 | |||
XM_017012729.1 | 12440 | UTR 3 | XP_016868218.1 | |||
XM_017012730.1 | 12440 | UTR 3 | XP_016868219.1 | |||
XM_017012731.1 | 12440 | UTR 3 | XP_016868220.1 | |||
XM_017012732.1 | 12440 | UTR 3 | XP_016868221.1 | |||
XM_017012733.1 | 12440 | UTR 3 | XP_016868222.1 | |||
XM_017012734.1 | 12440 | UTR 3 | XP_016868223.1 | |||
XM_017012735.1 | 12440 | UTR 3 | XP_016868224.1 | |||
XM_017012736.1 | 12440 | UTR 3 | XP_016868225.1 | |||
XM_017012737.1 | 12440 | UTR 3 | XP_016868226.1 | |||
XM_017012738.1 | 12440 | UTR 3 | XP_016868227.1 | |||
XM_017012739.1 | 12440 | Intron | XP_016868228.1 | |||
XM_017012740.1 | 12440 | Intron | XP_016868229.1 |