Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACTCTCCTCCCGGGACCCTCCGG[G/T]TTCTCTGTCCGCCAAGAAGGTCCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616048 | ||||||||||||||||||||
Literature Links: |
APTR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APTR - Alu-mediated CDKN1A/p21 transcriptional regulator (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RSBN1L - round spermatid basic protein 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198467.2 | 157 | Missense Mutation | GGT,GTT | G,V 38 | NP_940869.2 |