Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606821 MIM: 616714 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COG5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
COG5 - component of oligomeric golgi complex 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161520.1 | 3401 | Intron | NP_001154992.1 | |||
NM_006348.3 | 3401 | UTR 3 | NP_006339.3 | |||
NM_181733.2 | 3401 | UTR 3 | NP_859422.2 |
HBP1 - HMG-box transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244262.1 | 3401 | Intron | NP_001231191.1 | |||
NM_012257.3 | 3401 | Intron | NP_036389.2 | |||
XM_005250266.2 | 3401 | Intron | XP_005250323.1 | |||
XM_005250267.2 | 3401 | Intron | XP_005250324.1 | |||
XM_017011967.1 | 3401 | Intron | XP_016867456.1 |