Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606868 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIPK2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HIPK2 - homeodomain interacting protein kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113239.2 | 14222 | UTR 3 | NP_001106710.1 | |||
NM_022740.4 | 14222 | UTR 3 | NP_073577.3 | |||
XM_006715935.2 | 14222 | UTR 3 | XP_006715998.1 | |||
XM_011516077.2 | 14222 | UTR 3 | XP_011514379.1 | |||
XM_011516078.2 | 14222 | UTR 3 | XP_011514380.1 | |||
XM_011516079.2 | 14222 | UTR 3 | XP_011514381.1 | |||
XM_011516080.2 | 14222 | UTR 3 | XP_011514382.1 | |||
XM_011516081.2 | 14222 | UTR 3 | XP_011514383.1 | |||
XM_017012069.1 | 14222 | UTR 3 | XP_016867558.1 |