Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGCGGCGCTATCATCATCTGGC[A/G]CTTTGCTGGCAATTTCGAGAGAACC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615203 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RHBDD2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
RHBDD2 - rhomboid domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040456.1 | 297 | Missense Mutation | CAC,CGC | H,R 85 | NP_001035546.1 | |
NM_001040457.1 | 297 | UTR 5 | NP_001035547.1 | |||
XM_005250511.4 | 297 | UTR 5 | XP_005250568.1 |
Set Membership: |
HapMap |