Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609110 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO43 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FBXO43 - F-box protein 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029860.3 | Intron | NP_001025031.2 | ||||
XM_011516987.2 | Intron | XP_011515289.1 | ||||
XM_011516989.2 | Intron | XP_011515291.1 | ||||
XM_011516990.1 | Intron | XP_011515292.1 | ||||
XM_011516992.2 | Intron | XP_011515294.1 | ||||
XM_017013358.1 | Intron | XP_016868847.1 |
Set Membership: |
HapMap |