Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGAGTATCCATCACCCGCAGCA[A/G]CCGCTCAGGGAACACCATCAAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602019 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105375744 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105375744 - uncharacterized LOC105375744 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SQLE - squalene epoxidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003129.3 | 596 | UTR 5 | NP_003120.2 | |||
XM_011517246.2 | 596 | Intron | XP_011515548.1 |