Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614606 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOCAD PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FOCAD - focadhesin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017794.4 | Intron | NP_060264.4 | ||||
XM_005251494.3 | Intron | XP_005251551.2 | ||||
XM_011517945.2 | Intron | XP_011516247.1 | ||||
XM_017014852.1 | Intron | XP_016870341.1 | ||||
XM_017014853.1 | Intron | XP_016870342.1 | ||||
XM_017014854.1 | Intron | XP_016870343.1 | ||||
XM_017014855.1 | Intron | XP_016870344.1 | ||||
XM_017014856.1 | Intron | XP_016870345.1 | ||||
XM_017014857.1 | Intron | XP_016870346.1 | ||||
XM_017014858.1 | Intron | XP_016870347.1 | ||||
XM_017014859.1 | Intron | XP_016870348.1 |
FOCAD-AS1 - FOCAD antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |