Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610354 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RIC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RIC1 - RIC1 homolog, RAB6A GEF complex partner 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135920.2 | Intron | NP_001129392.2 | ||||
NM_001206557.1 | Intron | NP_001193486.1 | ||||
NM_020829.3 | Intron | NP_065880.2 | ||||
XM_005251523.3 | Intron | XP_005251580.1 | ||||
XM_011517967.2 | Intron | XP_011516269.1 | ||||
XM_011517968.2 | Intron | XP_011516270.1 | ||||
XM_017014934.1 | Intron | XP_016870423.1 | ||||
XM_017014935.1 | Intron | XP_016870424.1 | ||||
XM_017014936.1 | Intron | XP_016870425.1 |