Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCAGTTTCTCTCATTGGCATGAG[C/G]CTCACCCAGCAGCACTTCGGGTAGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604582 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKAP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AKAP2 - A-kinase anchoring protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004065.4 | Intron | NP_001004065.2 | ||||
NM_001136562.2 | Intron | NP_001130034.1 | ||||
NM_001198656.1 | Intron | NP_001185585.1 |
PALM2-AKAP2 - PALM2-AKAP2 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007203.4 | Intron | NP_009134.1 | ||||
NM_147150.2 | Intron | NP_671492.1 |
Set Membership: |
HapMap |