Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGTACTGAGCACAAAGTCTGACT[C/T]TACCAAGTACCAGCTGTGTGACTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616774 | ||||||||||||||||||||
Literature Links: |
MIGA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIGA2 - mitoguardin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005252282.1 | Intron | XP_005252339.1 | ||||
XM_011519117.1 | Intron | XP_011517419.1 | ||||
XM_011519118.1 | Intron | XP_011517420.1 |