Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600469 MIM: 611153 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCBP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NCBP1 - nuclear cap binding protein subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002486.4 | Intron | NP_002477.1 | ||||
XM_011518715.2 | Intron | XP_011517017.1 | ||||
XM_017014735.1 | Intron | XP_016870224.1 | ||||
XM_017014736.1 | Intron | XP_016870225.1 | ||||
XM_017014737.1 | Intron | XP_016870226.1 | ||||
XM_017014738.1 | Intron | XP_016870227.1 |
XPA - XPA, DNA damage recognition and repair factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000380.3 | Intron | NP_000371.1 | ||||
XM_006717278.1 | Intron | XP_006717341.1 | ||||
XM_017015114.1 | Intron | XP_016870603.1 |
Set Membership: |
HapMap |