Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605084 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MICU1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MICU1 - mitochondrial calcium uptake 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195518.1 | Intron | NP_001182447.1 | ||||
NM_001195519.1 | Intron | NP_001182448.1 | ||||
NM_006077.3 | Intron | NP_006068.2 | ||||
XM_005269383.2 | Intron | XP_005269440.1 | ||||
XM_005269386.2 | Intron | XP_005269443.1 | ||||
XM_011539119.1 | Intron | XP_011537421.1 |
Set Membership: |
HapMap |