Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCATGGCAATATTGCCCTAAAGCT[A/G]CCTAACTGTGACTATGGAAATTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 135630 | ||||||||||||||||||||
Literature Links: |
ITGB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ITGB1 - integrin subunit beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002211.3 | 2598 | UTR 3 | NP_002202.2 | |||
NM_033668.2 | 2598 | UTR 3 | NP_391988.1 | |||
NM_133376.2 | 2598 | UTR 3 | NP_596867.1 |
SNORA86 - small nucleolar RNA, H/ACA box 86 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |