Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCTGCAGGAGTTATGTTACTTTG[C/T]CACTTCTTCACGGACCAGTTTCAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606827 | ||||||||||||||||||||
Literature Links: |
PLXDC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLXDC2 - plexin domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282736.1 | 919 | Silent Mutation | TGC,TGT | C,C 18 | NP_001269665.1 | |
NM_032812.8 | 919 | Silent Mutation | TGC,TGT | C,C 18 | NP_116201.7 | |
XM_011519750.2 | 919 | Silent Mutation | TGC,TGT | C,C 18 | XP_011518052.1 |