Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCATTTCTGGGCACCATGGGCTC[C/T]ACAGTGTGCACAGATGAACGAAGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612754 | ||||||||||||||||||||
Literature Links: |
GLRX3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GLRX3 - glutaredoxin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199868.1 | 143 | Missense Mutation | CCA,CTA | P,L 44 | NP_001186797.1 | |
NM_001321980.1 | 143 | Intron | NP_001308909.1 | |||
NM_006541.4 | 143 | Intron | NP_006532.2 | |||
XM_017015474.1 | 143 | UTR 5 | XP_016870963.1 |