Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614460 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP47 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
USP47 - ubiquitin specific peptidase 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282659.1 | Intron | NP_001269588.1 | ||||
NM_017944.3 | Intron | NP_060414.3 | ||||
XM_005252997.1 | Intron | XP_005253054.1 | ||||
XM_011520196.1 | Intron | XP_011518498.1 | ||||
XM_017017953.1 | Intron | XP_016873442.1 | ||||
XM_017017954.1 | Intron | XP_016873443.1 |
Set Membership: |
HapMap |