Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCACTAGAGGGCATGCTCCTCCTA[C/G]CAACTGAAATAGTTGTAGGAATTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605306 | ||||||||||||||||||||
Literature Links: |
CLEC4A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLEC4A - C-type lectin domain family 4 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016184.3 | Intron | NP_057268.1 | ||||
NM_194447.2 | Intron | NP_919429.2 | ||||
NM_194448.2 | Intron | NP_919430.1 | ||||
NM_194450.2 | Intron | NP_919432.1 |