Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCCTTCCCTTCCTTTCTCCTTC[C/T]TTCCTTCCTTCCTCCCTCCCTCCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610086 | ||||||||||||||||||||
Literature Links: |
PRMT8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRMT8 - protein arginine methyltransferase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256536.1 | Intron | NP_001243465.1 | ||||
NM_019854.4 | Intron | NP_062828.3 | ||||
XM_017019644.1 | Intron | XP_016875133.1 | ||||
XM_017019645.1 | Intron | XP_016875134.1 |