Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612492 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP30 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
USP30 - ubiquitin specific peptidase 30 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301175.1 | Intron | NP_001288104.1 | ||||
NM_032663.4 | Intron | NP_116052.2 | ||||
XM_005253962.3 | Intron | XP_005254019.1 | ||||
XM_005253965.4 | Intron | XP_005254022.1 | ||||
XM_006719653.3 | Intron | XP_006719716.1 | ||||
XM_011538894.2 | Intron | XP_011537196.1 | ||||
XM_017020048.1 | Intron | XP_016875537.1 | ||||
XM_017020049.1 | Intron | XP_016875538.1 | ||||
XM_017020050.1 | Intron | XP_016875539.1 | ||||
XM_017020051.1 | Intron | XP_016875540.1 | ||||
XM_017020052.1 | Intron | XP_016875541.1 | ||||
XM_017020053.1 | Intron | XP_016875542.1 | ||||
XM_017020054.1 | Intron | XP_016875543.1 |
Set Membership: |
HapMap |