Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGCAGAGAATGGAAAGTCAAAT[C/T]TCCTGAATTGCTATGTGTCTGGGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 109700 MIM: 614661 | ||||||||||||||||||||
Literature Links: |
B2M PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
B2M - beta-2-microglobulin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004048.2 | 179 | Missense Mutation | CTC,TTC | L,F 42 | NP_004039.1 | |
XM_005254549.3 | 179 | Missense Mutation | CTC,TTC | L,F 42 | XP_005254606.1 |
PATL2 - PAT1 homolog 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |