Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604184 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGDCC3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IGDCC3 - immunoglobulin superfamily DCC subclass member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004884.3 | 2817 | UTR 3 | NP_004875.2 | |||
XM_011522241.1 | 2817 | UTR 3 | XP_011520543.1 | |||
XM_011522243.1 | 2817 | UTR 3 | XP_011520545.1 | |||
XM_011522244.1 | 2817 | UTR 3 | XP_011520546.1 | |||
XM_011522245.2 | 2817 | Intron | XP_011520547.1 |