Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTTTATTTCTATCCACAGGCACC[C/T]TAAGGATGTGATCCTACTGACAGCC
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 606881 | ||||||||||||||||||||||||||
Literature Links: |
FHOD1 PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
FHOD1 - formin homology 2 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318202.1 | 528 | Intron | NP_001305131.1 | |||
NM_013241.2 | 528 | Intron | NP_037373.2 | |||
XM_006721180.1 | 528 | Intron | XP_006721243.1 | |||
XM_011523043.2 | 528 | Intron | XP_011521345.1 | |||
XM_011523044.1 | 528 | Intron | XP_011521346.1 | |||
XM_011523045.2 | 528 | Intron | XP_011521347.1 |
LOC105369155 - uncharacterized LOC105369155 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC29 - leucine rich repeat containing 29 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM208 - transmembrane protein 208 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318217.1 | 528 | Missense Mutation | CCT,CTT | P,L 32 | NP_001305146.1 | |
NM_014187.3 | 528 | Missense Mutation | CCT,CTT | P,L 102 | NP_054906.2 |