Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611510 MIM: 605813 MIM: 609119 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CENPT PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CENPT - centromere protein T | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUTF2 - nuclear transport factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322038.1 | Intron | NP_001308967.1 | ||||
NM_001322039.1 | Intron | NP_001308968.1 | ||||
NM_001322040.1 | Intron | NP_001308969.1 | ||||
NM_001322041.1 | Intron | NP_001308970.1 | ||||
NM_005796.2 | Intron | NP_005787.1 |
THAP11 - THAP domain containing 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |