Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGAGAGATGCTCTCAGCACTGGAT[G/T]GCAAACCCGGGAAGATGGCGTCGGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616951 MIM: 604632 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTSS1L PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MTSS1L - metastasis suppressor 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VAC14 - Vac14, PIKFYVE complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018052.3 | 4407 | Intron | NP_060522.3 | |||
XM_005256038.3 | 4407 | UTR 3 | XP_005256095.1 | |||
XM_005256041.3 | 4407 | Intron | XP_005256098.2 | |||
XM_011523223.2 | 4407 | Intron | XP_011521525.1 | |||
XM_011523224.2 | 4407 | Intron | XP_011521526.1 | |||
XM_011523225.2 | 4407 | Intron | XP_011521527.1 | |||
XM_017023441.1 | 4407 | Intron | XP_016878930.1 | |||
XM_017023442.1 | 4407 | Intron | XP_016878931.1 |
Set Membership: |
HapMap |