Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616951 MIM: 604632 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTSS1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTSS1L - metastasis suppressor 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VAC14 - Vac14, PIKFYVE complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018052.3 | 2373 | Silent Mutation | NP_060522.3 | |||
XM_005256038.3 | 2373 | Silent Mutation | XP_005256095.1 | |||
XM_005256041.3 | 2373 | Intron | XP_005256098.2 | |||
XM_011523223.2 | 2373 | Intron | XP_011521525.1 | |||
XM_011523224.2 | 2373 | Intron | XP_011521526.1 | |||
XM_011523225.2 | 2373 | Intron | XP_011521527.1 | |||
XM_017023441.1 | 2373 | Intron | XP_016878930.1 | |||
XM_017023442.1 | 2373 | Intron | XP_016878931.1 |