Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGAGGCGACATCGCTCCACTGGA[A/G]GCCAAGCCACAGCCTCGGGCGGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114761 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CA5A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CA5A - carbonic anhydrase 5A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001739.1 | Intron | NP_001730.1 | ||||
XM_005256134.4 | Intron | XP_005256191.1 | ||||
XM_011523309.2 | Intron | XP_011521611.1 | ||||
XM_011523310.2 | Intron | XP_011521612.1 | ||||
XM_017023646.1 | Intron | XP_016879135.1 |