Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTCACAAGAGACAGTGTGTCCAT[G/T]AGAAAATGCTTCCGCAGCTCCCCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 182115 | ||||||||||||||||||||
Literature Links: |
CYTH1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYTH1 - cytohesin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001292018.1 | Intron | NP_001278947.1 | ||||
NM_001292019.1 | Intron | NP_001278948.1 | ||||
NM_004762.3 | Intron | NP_004753.1 | ||||
NM_017456.3 | Intron | NP_059430.2 | ||||
XM_011525475.2 | Intron | XP_011523777.1 | ||||
XM_011525476.2 | Intron | XP_011523778.1 | ||||
XM_011525477.1 | Intron | XP_011523779.1 | ||||
XM_011525478.1 | Intron | XP_011523780.1 | ||||
XM_017025336.1 | Intron | XP_016880825.1 | ||||
XM_017025337.1 | Intron | XP_016880826.1 | ||||
XM_017025338.1 | Intron | XP_016880827.1 |